missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Casein kinase I isoform epsilon Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase I isoform epsilon, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001276841.1 |
---|---|
Gene ID (Entrez) | 102800317 |
Name | Casein kinase I isoform epsilon Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16014211
|
enQuireBio™
QP8358-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034211
|
enQuireBio™
QP8358-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024211
|
enQuireBio™
QP8358-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044211
|
enQuireBio™
QP8358-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054211
|
enQuireBio™
QP8358-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064211
|
enQuireBio™
QP8358-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001276841.1 | |
Casein kinase I isoform epsilon Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
102800317 | |
Research Use Only | |
CSNK1E | |
Recombinant Protein | |
E. coli | |
SGSFGDIYLG ANIASGEEVA IKLECVKTKH PQLHIESKFY KMMQGGVGIP SIKWCGAEGD YNVMVMELLG PSLEDLFNFC SRKFSLKTVL LLADQMISRI EYIHSKNFIH RDVKPDNFLM GLGKKGNLVY IIDFGLAKKY RDARTHQHIP YRENKNLTGT ARYASINTHL GIEQSRRDDL ESLGYVLMYF NLGSLPWQGL KAATKRQKYE RISEK | |
Greater than 90% as determined by SDS-PAGE. |