missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Caspase-14 / CASP14 Protein
A DNA sequence encoding the Homo sapiens (Human) Caspase-14 / CASP14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5761-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036246.1 | |
Caspase-14 / CASP14 Protein | |
Research Use Only | |
CASP14 | |
Recombinant Protein | |
E. coli | |
KDSPQTIPTY TDALHVYSTV EGYIAYRHDQ KGSCFIQTLV DVFTKRKGHI LELLTEVTRR MAEAELVQEG KARKTNPEIQ STLRKRLY | |
Greater than 90% as determined by SDS-PAGE. |
23581 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |