missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCDC25 Protein

A cDNA sequence encoding the CCDC25 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11289-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986728

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CCDC25 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CCDC25 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMVFYFTS SSVNSSAYTI YMGKDKYENE DLIKHGWPED IWFHVDKLSS AHVYLRLHKG ENIEDIPKEV LMDCAHLVKA NSIQGCKMNN VNVVYTPWSN LKKTADMDVG QIGFHRQKDV KIVTVEKKVN EILNRLEKTK VERFPDLAAE KECRDREERN EKKAQIQEMK KREKEEMKKK REMDELRSYS SLMKVENMSS NQDGNDSDEF M
Videos
SDS
Documents
Certificates