missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCDC90B Protein
A cDNA sequence encoding the CCDC90B was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 60492 |
---|---|
Name | CCDC90B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CCDC90B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15986738
|
enQuireBio™
QP11292-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976738
|
enQuireBio™
QP11292-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966738
|
enQuireBio™
QP11292-100UG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
60492 | |
Research Use Only | |
CCDC90B | |
Human | |
His | |
CCDC90B protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
CCDC90B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGYDRRPV DITPLEQRKL TFDTHALVQD LETHGFDKTQ AETIVSALTA LSNVSLDTIY KEMVTQAQQE ITVQQLMAHL DAIRKDMVIL EKSEFANLRA ENEKMKIELD QVKQQLMHET SRIRADNKLD INLERSRVTD MFTDQEKQLM ETTTEFTKKD TQTKSIISET SNKIDAEIAS LKTLMESNKL E | |
Greater than 90% as determined by SDS-PAGE. |