missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCDC90B Protein
A cDNA sequence encoding the CCDC90B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11292-2ug
Additional Details : Weight : 0.01000kg
Specifications
60492 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGYDRRPV DITPLEQRKL TFDTHALVQD LETHGFDKTQ AETIVSALTA LSNVSLDTIY KEMVTQAQQE ITVQQLMAHL DAIRKDMVIL EKSEFANLRA ENEKMKIELD QVKQQLMHET SRIRADNKLD INLERSRVTD MFTDQEKQLM ETTTEFTKKD TQTKSIISET SNKIDAEIAS LKTLMESNKL E | |
Greater than 90% as determined by SDS-PAGE. |
CCDC90B Protein | |
Research Use Only | |
CCDC90B | |
Human | |
His | |
CCDC90B protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |