missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL21 / 6Ckine Protein
Recombinant Protein
2490.00 € - 4060.00 €
Specifications
Accession Number | NP_002980.1 |
---|---|
Gene ID (Entrez) | 6366 |
Name | CCL21 / 6Ckine Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985739
|
enQuireBio™
QP5229-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975739
|
enQuireBio™
QP5229-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_002980.1 | |
CCL21 / 6Ckine Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
SDGGAQDCCL KYSQRKIPAK VVRSYRKQEP SLGCSIPAIL FLPRKRSQAE LCADPKELWV QQLMQHLDKT PSPQKPAQGC RKDRGASKTG KKGKGSKGCK RTERSQTPKG P | |
Greater than 95% as determined by SDS-PAGE. |
6366 | |
Research Use Only | |
CCL21 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |