missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL8 / MCP-2 Protein
Recombinant Protein
Brand: enQuireBio™ QP5251-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005614.2 | |
CCL8 / MCP-2 Protein | |
Research Use Only | |
CCL8 | |
Recombinant Protein | |
E. coli | |
QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP | |
Greater than 95% as determined by SDS-PAGE. |
6355 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CCL8 / MCP-2 induces Human THP-1 chemotaxis (significanty different from control at Human CCL8 / MCP-2 concentrations greater than 100 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |