missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL8 / MCP-2 Protein
Recombinant Protein
2490.00 € - 4060.00 €
Specifications
Accession Number | NP_005614.2 |
---|---|
Gene ID (Entrez) | 6355 |
Name | CCL8 / MCP-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985769
|
enQuireBio™
QP5251-500UG |
500 μg |
2490.00 €
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975769
|
enQuireBio™
QP5251-1MG |
1 mg |
4060.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_005614.2 | |
CCL8 / MCP-2 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CCL8 / MCP-2 induces Human THP-1 chemotaxis (significanty different from control at Human CCL8 / MCP-2 concentrations greater than 100 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6355 | |
Research Use Only | |
CCL8 | |
Recombinant Protein | |
E. coli | |
QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP | |
Greater than 95% as determined by SDS-PAGE. |