missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCNB2 Protein
A cDNA sequence encoding the CCNB2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 9133 |
---|---|
Name | CCNB2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CCNB2 |
Product Code | Brand | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
15976748
|
enQuireBio™
QP11295-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956748
|
enQuireBio™
QP11295-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966748
|
enQuireBio™
QP11295-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Spezifikation
9133 | |
Research Use Only | |
CCNB2 | |
Human | |
His | |
The CCNB2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT and 50% glycerol. |
CCNB2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMALLRR PTVSSDLENI DTGVNSKVKS HVTIRRTVLE EIGNRVTTRA AQVAKKAQNT KVPVQPTKTT NVNKQLKPTA SVKPVQMEKL APKGPSPTPE DVSMKEENLC QAFSDALLCK IEDIDNEDWE NPQLCSDYVK DIYQYLRQLE VLQSINPHFL DGRDINGRMR AILVDWLVQV HSKFRLLQET LYMCVGIMDR FLQVQPVSRK KLQLVGITAL LLASKYEEMF SPNIEDFVYI TDNAYTSSQI REMETLILKE LKFELGRPLP LHFLRRASKA GEVDVEQHTL AKYLMELTLI DYDMVHYHPS KVAAAASCLS QKVLGQGKWN LKQQYYTGYT ENEVLEVMQH MAKNVVKVNE NLTKFIAIKN KYASSKLLKI SMIPQLNSKA VKDLASPLIG RSc | |
Greater than 90% as determined by SDS-PAGE. |