missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD164 Protein

A cDNA sequence encoding the CD164 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11305-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986778

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

8763
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
CD164
Human
His
CD164 protein solution (0.5 mg/ml) contains 10% glycerol and Phosphate buffered saline (pH 7.4).
CD164 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
ADPDKNTTQH PNVTTLAPIS NVTSAPVTSL PLVTTPAPET CEGRNSCVSC FNVSVVNTTC FWIECKDESY CSHNSTVSDC QVGNTTDFCS VSTATPVPTA NSTAKPTVQP SPSTTSKTVT TSGTTNNTVT PTSQPVRKST FDHHHHHH
Videos
SDS
Documents
Certificates