missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD164 Protein
A cDNA sequence encoding the CD164 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 8763 |
---|---|
Name | CD164 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD164 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906788
|
enQuireBio™
QP11305-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986778
|
enQuireBio™
QP11305-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996778
|
enQuireBio™
QP11305-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
8763 | |
Research Use Only | |
CD164 | |
Human | |
His | |
CD164 protein solution (0.5 mg/ml) contains 10% glycerol and Phosphate buffered saline (pH 7.4). |
CD164 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPDKNTTQH PNVTTLAPIS NVTSAPVTSL PLVTTPAPET CEGRNSCVSC FNVSVVNTTC FWIECKDESY CSHNSTVSDC QVGNTTDFCS VSTATPVPTA NSTAKPTVQP SPSTTSKTVT TSGTTNNTVT PTSQPVRKST FDHHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |