missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD1E Protein

A cDNA sequence encoding the CD1E was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11311-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986798

  • 131.00 € / 5µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

913
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEEQLSFR MLQTSSFANH SWAHSEGSGW LGDLQTHGWD TVLGTIRFLK PWSHGNFSKQ ELKNLQSLFQ LYFHSFIQIV QASAGQFQLE YPFEIQILAG CRMNAPQIFL NMAYQGSDFL SFQGISWEPS PGAGIRAQNI CKVLNRYLDI KEILQSLLGH TCPRFLAGLM EAGESELKRK VKPEAWLSCG PSPGPGRLQL VCHVSGFYPK PVWVMWMRGE QEQRGTQRGD VLPNADETWY LRATLDVAAG EAAGLSCRVK HSSLGGHDLI IHWGGYS
Greater than 85.0% as determined by SDS-PAGE.
CD1E Protein
Research Use Only
CD1E
Human
His
CD1E protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers