missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD247 Protein
A cDNA sequence encoding the CD247 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11317-2ug
Additional Details : Weight : 0.01000kg
Specifications
919 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRVKFSRS ADAPAYQQGQ NQLYNELNLG RREEYDVLDK RRGRDPEMGG KPQRRKNPQE GLYNELQKDK MAEAYSEIGM KGERRRGKGH DGLYQGLSTA TKDTYDALHM QALPPR | |
Greater than 90% as determined by SDS-PAGE. |
CD247 Protein | |
Research Use Only | |
CD247 | |
Human | |
His | |
The CD247 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |