missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD274 Protein

A cDNA sequence encoding the CD274 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11319-HIS-INSECT-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986838

Quantity Price
1 1511.71 € / 100µg
3 1421.92 € / 100µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

29126
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CD274
Human
His
The CD274 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol.
CD274 Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
MGSSHHHHHH SSGLVPRGSH MGSHMFTVTV PKDLYVVEYG SNMTIECKFP VEKQLDLAAL IVYWEMEDKN IIQFVHGEED LKVQHSSYRQ RARLLKDQLS LGNAALQITD VKLQDAGVYR CMISYGGADY KRITVKVNAP YNKINQRILV VDPVTSEHEL TCQAEGYPKA EVIWTSSDHQ VLSGKTTTTN SKREEKLFNV TSTLRINTTT NEIFYCTFRR LDPEENHTAE LVIPELPLAH PPNER
Videos
SDS
Documents
Certificates