missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD37 Protein
A cDNA sequence encoding the CD37 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 951 |
---|---|
Name | CD37 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD37 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15956888
|
enQuireBio™
QP11330-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946888
|
enQuireBio™
QP11330-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936888
|
enQuireBio™
QP11330-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
951 | |
Research Use Only | |
CD37 | |
Human | |
His | |
CD37 protein solution (1 mg/ml) containing Phosphate buffer saline, (pH 7.4) and 10% glycerol. |
CD37 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQRAQLER SLRDVVEKTI QKYGTNPEET AAEESWDYVQ FQLRCCGWHY PQDWFQVLIL RGNGSEAHRV PCSCYNLSAT NDSTILDKVI LPQLSRLGHL ARSRHSADIC AVPAESHIYR EGCAQGLQKW LHNN | |
Greater than 90.0% as determined by SDS-PAGE. |