missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD38 Protein
A cDNA sequence encoding the CD38 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11331-2ug
Additional Details : Weight : 0.01000kg
Specifications
952 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPEFVPRWR QQWSGPGTTK RFPETVLARC VKYTEIHPEM RHVDCQSVWD AFKGAFISKH PCNITEEDYQ PLMKLGTQTV PCNKILLWSR IKDLAHQFTQ VQRDMFTLED TLLGYLADDL TWCGEFNTSK INYQSCPDWR KDCSNNPVSV FWKTVSRRFA EAACDVVHVM LNGSRSKIFD KNSTFGSVEV HNLQPEKVQT LEAWVIHGGR EDSRDLCQDP TIKELESIIS KRNIQFSCKN IYRPDKFLQC VKNPEDSSCT SEIHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
CD38 Protein | |
Research Use Only | |
CD38 | |
Human | |
His | |
CD38 protein solution (1 mg/ml) contains 100mM NaCl and 10% glycerol. |