missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD40 Protein
A cDNA sequence encoding the CD40 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 958 |
---|---|
Name | CD40 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD40 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976898
|
enQuireBio™
QP11333-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956898
|
enQuireBio™
QP11333-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966898
|
enQuireBio™
QP11333-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
958 | |
Research Use Only | |
CD40 | |
Human | |
His | |
CD40 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
CD40 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEPPTACR EKQYLINSQC CSLCQPGQKL VSDCTEFTET ECLPCGESEF LDTWNRETHC HQHKYCDPNL GLRVQQKGTS ETDTICTCEE GWHCTSEACE SCVLHRSCSP GFGVKQIATG VSDTICEPCP VGFFSNVSSA FEKCHPWTSC ETKDLVVQQA GTNKTDVVCG PQDRLR | |
Greater than 90.0% as determined by SDS-PAGE. |