missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD47 Protein

A cDNA sequence encoding the CD47 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11336-HIS-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986908

Quantity Price
1 114.77 € / 2µg
10 107.33 € / 2µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

961
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
CD47
Human
His
CD47 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
CD47 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
QLLFNKTKSV EFTFCNDTVV IPCFVTNMEA QNTTEVYVKW KFKGRDIYTF DGALNKSTVP TDFSSAKIEV SQLLKGDASL KMDKSDAVSH TGNYTCEVTE LTREGETIIE LKYRVVSWFS PNELEPKSCD KTHTCPPCPA PELLGGPSVF LFPPKPKDTL MISRTPEVTC VVVDVSHEDPEVKFNWYVDG VEVHNAKTKP REEQYNSTYR VVSVLTVLHQ DWLNGKEYKC KVSNKALPAP IEKTISKAKG QPREPQVYTL PPSRDELTKN QVSLTCLVKG FYPSDIAVEW ESNGQPENNY KTTPPVLDSD GSFFLYSKLT VDKSRWQQGN VFSCSVMHEA LHNHYTQKSL SLSPGKHHHH HH
Videos
SDS
Documents
Certificates