missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD48 Protein
A cDNA sequence encoding the CD48 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Gene ID (Entrez) | 962 |
---|---|
Name | CD48 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD48 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15926918
|
enQuireBio™
QP11337-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906918
|
enQuireBio™
QP11337-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916918
|
enQuireBio™
QP11337-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
962 | |
Research Use Only | |
CD48 | |
Human | |
His | |
NECTIN2 protein solution (1.0 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
CD48 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPQGHLVHM TVVSGSNVTL NISESLPENY KQLTWFYTFD QKIVEWDSRK SKYFESKFKG RVRLDPQSGA LYISKVQKED NSTYIMRVLK KTGNEQEWKI KLQVLDPVPK PVIKIEKIED MDDNCYLKLS CVIPGESVNY TWYGDKRPFP KELQNSVLET TLMPHNYSRC YTCQVSNSVSSKNGTVCLSP PCTLARSHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |