missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD68 Protein

A cDNA sequence encoding the CD68 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11343-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986938

Quantity Price
1 228.51 € / 2µg
6 214.69 € / 2µg
Estimated Shipment: 26-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

968
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
MRLAVLFSGA LLGLLAAQGT GNDCPHKKSA TLLPSFTVTP TVTESTGTTS HRTTKSHKTT THRTTTTGTT SHGPTTATHN PTTTSHGNVT VHPTSNSTAT SQGPSTATHS PATTSHGNAT VHPTSNSTAT SPGFTSSAHP EPPPPSPSPS PTSKETIGDY TWTNGSQPCV HLQAQIQIRV MYTTQGGGEA WGISVLNPNK TKVQGSCEGA HPHLLLSFPY GHLSFGFMQD LQQKVVYLSY MAVEYNVSFP HAAQWTFSAQ NASLRDLQAP LGQSFSCSNS SIILSPAVHL DLLSLRLQAA QLPHTGVFGQ SFSCPSDRSI LLPLIIGLIL LGLLALVLIA FCIIRRRPSA YQAL
CD68 Protein
Research Use Only
CD68
Human
Untagged
CD68 at 100microg/1ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA, 20% Glycerol.
Videos
SDS
Documents
Certificates