missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD79A Protein

A cDNA sequence encoding the CD79A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11347-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15986958

Quantity Price
1 114.77 € / 2µg
10 107.33 € / 2µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

973
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
CD79A
Human
His
CD79A protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
CD79A Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
LWMHKVPASL MVSLGEDAHF QCPHNSSNNA NVTWWRVLHG NYTWPPEFLG PGEDPNGTLI IQNVNKSHGG IYVCRVQEGN ESYQQSCGTY LRVRQPPPRP FLDMGEGTKN RLEHHHHHH
Videos
SDS
Documents
Certificates