missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD84 Protein
A cDNA sequence encoding the CD84 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11352-HIS-EC-1mg
Additional Details : Weight : 0.01000kg
Specifications
8832 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMKDSEI FTVNGILGES VTFPVNIQEP RQVKIIAWTS KTSVAYVTPG DSETAPVVTV THRNYYERIH ALGPNYNLVI SDLRMEDAGD YKADINTQAD PYTTTKRYNL QIYRRLGKPK ITQSLMASVN STCNVTLTCS VEKEEKNVTY NWSPLGEEGN VLQIFQTPED QELTYTCTAQ NPVSNNSDSI SARQLCADIA MGFRTHHTG | |
Greater than 80% as determined by SDS-PAGE. |
CD84 Protein | |
Research Use Only | |
CD84 | |
Human | |
His | |
The CD84 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |