missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CDK2AP2 Protein
A cDNA sequence encoding the CDK2AP2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 10263 |
---|---|
Name | CDK2AP2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CDK2AP2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15927068
|
enQuireBio™
QP11374-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917068
|
enQuireBio™
QP11374-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907068
|
enQuireBio™
QP11374-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10263 | |
Research Use Only | |
CDK2AP2 | |
Human | |
His | |
CDK2AP2 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |
CDK2AP2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSYKPIA PAPSSTPGSS TPGPGTPVPT GSVPSPSGSV PGAGAPFRPL FNDFGPPSMG YVQAMKPPGA QGSQSTYTDL LSVIEEMGKE IRPTYAGSKS AMERLKRGII HARALVRECL AETERNART | |
Greater than 95.0% as determined by SDS-PAGE. |