missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEACAM1 / CD66a Protein
A DNA sequence encoding the Homo sapiens (Human) CEACAM1 / CD66a, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001020083.1 |
---|---|
Gene ID (Entrez) | 634 |
Name | CEACAM1 / CD66a Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987919
|
enQuireBio™
QP5811-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15907929
|
enQuireBio™
QP5811-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997919
|
enQuireBio™
QP5811-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15917929
|
enQuireBio™
QP5811-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15927929
|
enQuireBio™
QP5811-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15937929
|
enQuireBio™
QP5811-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001020083.1 | |
CEACAM1 / CD66a Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
634 | |
Research Use Only | |
CEACAM1 | |
Recombinant Protein | |
E. coli | |
QLTTESMPFN VAEGKEVLLL VHNLPQQLFG YSWYKGERVD GNRQIVGYAI GTQQATPGPA NSGRETIYPN ASLLIQNVTQ NDTGFYTLQV IKSDLVNEEA TGQFHVYPEL PKPSISSNNS NPVEDKDAVA FTCEPETQDT TYLWWINNQS LPVSPRLQLS NGNRTLTLLS VTRNDTGPYE CEIQNPVSAN RSDPVTLNVT YGPDTPTISP SDTYYRPGAN LSLSCYAASN PPAQYSWLIN GTFQQSTQEL FIPNITVNNS GSYTCHANNS VTGCNRTTVK TIIVTELSPV VAKPQIKASK TTVTGDKDSV NLTCSTNDTG ISIRWFFKNQ SLPSSERMKL SQGNTTLSIN PVKREDAGTY WCEVFNPISK NQSDPIMLNV NYNALPQENG LSPG | |
Greater than 90% as determined by SDS-PAGE. |