missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEACAM6 Protein
A DNA sequence encoding the Homo sapiens (Human) CEACAM6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5813-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002474.4 | |
CEACAM6 Protein | |
Research Use Only | |
CEACAM6 | |
Recombinant Protein | |
E. coli | |
KLTIESTPFN VAEGKEVLLL AHNLPQNRIG YSWYKGERVD GNSLIVGYVI GTQQATPGPA YSGRETIYPN ASLLIQNVTQ NDTGFYTLQV IKSDLVNEEA TGQFHVYPEL PKPSISSNNS NPVEDKDAVA FTCEPEVQNT TYLWWVNGQS LPVSPRLQLS NGNMTLTLLS VKRNDAGSYE CEIQNPASAN RSDPVTLNVL YGPDGPTISP SKANYRPGEN LNLSCHAASN PPAQYSWFIN GTFQQSTQEL FIPNITVNNS GSYMCQAHNS ATGLNRTTVT MITVSG | |
Greater than 90% as determined by SDS-PAGE. |
4680 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |