missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CENPU Protein

A cDNA sequence encoding the CENPU was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11392-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987118

Quantity Price
1 171.64 € / 20µg
10 161.27 € / 20µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CENPU Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CENPU protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea.
Greater than 80.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSTRRKVKS AEKISTQRHE VIRTTASSEL SEKPAESVTS KKTGPLSAQP SVEKENLAIE SQSKTQKKGK ISHDKRKKSR SKAIGSDTSD IVHIWCPEGM KTSDIKELNI VLPEFEKTHL EHQQRIESKV CKAAIATFYV NVKEQFIKML KESQMLTNLK RKNAKMISDI EKKRQRMIEV QDELLRLEPQ LKQLQTKYDE LKERKSSLRN AAYFLSNLKQ LYQDYSDVQA QEPNVKETYD SSSLPALLFK ARTLLGAESH LRNINHQLEK LLDQG
Videos
SDS
Documents
Certificates