missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centrosomal protein of 63 kDa Protein
A DNA sequence encoding the Homo sapiens (Human) Centrosomal protein of 63 kDa, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001035842.1 |
---|---|
Gene ID (Entrez) | 80254 |
Name | Centrosomal protein of 63 kDa Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051471
|
enQuireBio™
QP7898-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071471
|
enQuireBio™
QP7898-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061471
|
enQuireBio™
QP7898-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081471
|
enQuireBio™
QP7898-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091471
|
enQuireBio™
QP7898-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001481
|
enQuireBio™
QP7898-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001035842.1 | |
Centrosomal protein of 63 kDa Protein | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
80254 | |
Research Use Only | |
CEP63 | |
Recombinant Protein | |
E. coli | |
MEALLEGIQN RGHGGGFLTS CEAELQELMK QIDIMVAHKK SEWEGRTHAL ETCLKIREQE LKSLRSQLDV THKEVGMLHQ QVEEHEKIKQ EMTMEYKQEL KKLHEELCIL KRSYEKLQKK QMREFRGNTK NHREDRSEIE RLTAKIEEFR QKSLDWEKQR LIYQQQVSSL EAQRKALAEQ SEIIQAQLVN RKQKLESVEL SSQSEIQHLS SKLERANDTI CANELEIERL TMRVNDLVGT SMTVLQEQQQ KEEKLRESEK LLEALQEEKR ELKAALQSQE NLIHEARIQK EKLQEKVKAT NTQHAVEAIS LESVSATCKQ LSQELMEKYE ELKRMEAHNN EYKAEIKKLK EQILQGEQSY SSALEGMKME ISHLTQELHQ RDITIASTKG SSSDMEKRLR AEMQKAEDKA VEHKEILDQL ESLKLENRHL SEMVMKLELG LHECSLPVSP LGSIATRFLE EEELRSHHIL ERLDAHIEEL KRESEKTVRQ FTALK | |
Greater than 90% as determined by SDS-PAGE. |