missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHMP4A Protein
A cDNA sequence encoding the CHMP4A was constructed and used to recombinantly synthesize the protein.
131.00 € - 1595.00 €
Specifications
Name | CHMP4A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15977208
|
enQuireBio™
QP11421-1UG |
1 μg |
131.00 €
1µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997208
|
enQuireBio™
QP11421-5UG |
5 μg |
203.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987208
|
enQuireBio™
QP11421-50UG |
50 μg |
1595.00 €
50µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CHMP4A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CHMP4A solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 0.1mMPMSF, 1mM EDTA, 2mM DTT and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSRRRPEDGL GKAGPCVMRH HPPRSKAEVW RTLRGGGGRG ELAMSGLGRL FGKGKKEKGP TPEEAIQKLK ETEKILIKKQ EFLEQKIQQE LQTAKKYGTK NKRAALQALR RKKRFEQQLA QTDGTLSTLE FQREAIENAT TNAEVLRTME LAAQSMKKAY QDMDIDKVDE LMTDITEQQE VAQQISDAIS RPMGFGDDVD EDELLEELEE LEQEELAQEL LNVGDKEEEP SVKLPSVPST HLPAGPAPKV DEDEEALKQL AEWVS | |
Greater than 90% as determined by SDS-PAGE. |