missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CHMP4A Protein

A cDNA sequence encoding the CHMP4A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11421-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987208

Quantity Price
1 1386.59 € / 50µg
3 1303.39 € / 50µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CHMP4A Protein
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CHMP4A solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 0.1mMPMSF, 1mM EDTA, 2mM DTT and 50% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSRRRPEDGL GKAGPCVMRH HPPRSKAEVW RTLRGGGGRG ELAMSGLGRL FGKGKKEKGP TPEEAIQKLK ETEKILIKKQ EFLEQKIQQE LQTAKKYGTK NKRAALQALR RKKRFEQQLA QTDGTLSTLE FQREAIENAT TNAEVLRTME LAAQSMKKAY QDMDIDKVDE LMTDITEQQE VAQQISDAIS RPMGFGDDVD EDELLEELEE LEQEELAQEL LNVGDKEEEP SVKLPSVPST HLPAGPAPKV DEDEEALKQL AEWVS
Videos
SDS
Documents
Certificates