missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMT2 Protein
A cDNA sequence encoding the CKMT2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Especificaciones
Gene ID (Entrez) | 1160 |
---|---|
Name | CKMT2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CKMT2 |
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
15997258
|
enQuireBio™
QP11437-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987258
|
enQuireBio™
QP11437-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977258
|
enQuireBio™
QP11437-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Especificaciones
1160 | |
Research Use Only | |
CKMT2 | |
Human | |
His | |
The CKMT2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |
CKMT2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEVREQ PRLFPPSADY PDLRKHNNCM AECLTPAIYA KLRNKVTPNG YTLDQCIQTG VDNPGHPFIK TVGMVAGDEE SYEVFADLFD PVIKLRHNGY DPRVMKHTTD LDASKITQGQ FDEHYVLSSR VRTGRSIRGL SLPPACTRAE RREVENVAIT ALEGLKGDLA GRYYKLSEMT EQDQQRLIDD HFLFDKPVSP LLTCAGMARD WPDARGIWHN YDKTFLIWIN EEDHTRVISM EKGGNMKRVF ERFCRGLKEV ERLIQERGWE FMWNERLGYI LTCPSNLGTG LRAGVHVRIP KLSKDPRFSK ILENLRLQKR GTGGVDTAAV ADVYDISNID RIGRSEVELV QIVIDGVNYL VDCEKKLERG QDIKVPPPLP QFGKK | |
Greater than 90% as determined by SDS-PAGE. |