missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMT2 Protein
A cDNA sequence encoding the CKMT2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11437-20ug
Additional Details : Weight : 0.01000kg
Specifications
1160 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMEVREQ PRLFPPSADY PDLRKHNNCM AECLTPAIYA KLRNKVTPNG YTLDQCIQTG VDNPGHPFIK TVGMVAGDEE SYEVFADLFD PVIKLRHNGY DPRVMKHTTD LDASKITQGQ FDEHYVLSSR VRTGRSIRGL SLPPACTRAE RREVENVAIT ALEGLKGDLA GRYYKLSEMT EQDQQRLIDD HFLFDKPVSP LLTCAGMARD WPDARGIWHN YDKTFLIWIN EEDHTRVISM EKGGNMKRVF ERFCRGLKEV ERLIQERGWE FMWNERLGYI LTCPSNLGTG LRAGVHVRIP KLSKDPRFSK ILENLRLQKR GTGGVDTAAV ADVYDISNID RIGRSEVELV QIVIDGVNYL VDCEKKLERG QDIKVPPPLP QFGKK | |
Greater than 90% as determined by SDS-PAGE. |
CKMT2 Protein | |
Research Use Only | |
CKMT2 | |
Human | |
His | |
The CKMT2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT and 10% glycerol. |