missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Clathrin heavy chain 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Clathrin heavy chain 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5842-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001826.3 | |
Clathrin heavy chain 2 Protein | |
Research Use Only | |
CLTCL1 | |
Recombinant Protein | |
E. coli | |
LLVLSPRLDH TWTVSFFSKA GQLPLVKPYL RSVQSHNNKS VNEALNHLLT EEEDYQGLRA SIDAYDNFDN ISLAQQLEKH QLMEFRCIAA YLYKGNNWWA QSVELCKKDH LYKDAMQHAA ESRDAELAQK LLQWFLEEGK RECF | |
Greater than 90% as determined by SDS-PAGE. |
8218 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |