missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLIC2 Protein
A cDNA sequence encoding the CLIC2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | CLIC2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957298
|
enQuireBio™
QP11448-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947298
|
enQuireBio™
QP11448-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937298
|
enQuireBio™
QP11448-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CLIC2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CLIC2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol 0.1M NaCl and 1mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGLRPGTQV DPEIELFVKA GSDGESIGNC PFCQRLFMIL WLKGVKFNVT TVDMTRKPEE LKDLAPGTNP PFLVYNKELK TDFIKIEEFL EQTLAPPRYP HLSPKYKESF DVGCNLFAKF SAYIKNTQKE ANKNFEKSLL KEFKRLDDYL NTPLLDEIDPDSAEEPPVSR RLFLDGDQLT LADCSLLPKL NIIKVAAKKY RDFDIPAEFS GVWRYLHNAY AREEFTHTCP EDKEIENTYA NVAKQKS | |
Greater than 95.0% as determined by SDS-PAGE. |