missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CLIC2 Protein
A cDNA sequence encoding the CLIC2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11448-5ug
Additional Details : Weight : 0.01000kg
Specifications
CLIC2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The CLIC2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol 0.1M NaCl and 1mM DTT. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGLRPGTQV DPEIELFVKA GSDGESIGNC PFCQRLFMIL WLKGVKFNVT TVDMTRKPEE LKDLAPGTNP PFLVYNKELK TDFIKIEEFL EQTLAPPRYP HLSPKYKESF DVGCNLFAKF SAYIKNTQKE ANKNFEKSLL KEFKRLDDYL NTPLLDEIDPDSAEEPPVSR RLFLDGDQLT LADCSLLPKL NIIKVAAKKY RDFDIPAEFS GVWRYLHNAY AREEFTHTCP EDKEIENTYA NVAKQKS |