missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CLMP Protein

A cDNA sequence encoding the CLMP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11449-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987298

Quantity Price
1 114.77 € / 5µg
10 107.33 € / 5µg
Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CLMP Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CLMP solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MASMTGGQQM GRGSHMTHTE IKRVAEEKVT LPCHHQLGLP EKDTLDIEWL LTDNEGNQKV VITYSSRHVY NNLTEEQKGR VAFASNFLAG DASLQIEPLK PSDEGRYTCK VKNSGRYVWS HVILKVLVRP SKPKCELEGE LTEGSDLTLQ CESSSGTEPI VYYWQRIREK EGEDERLPPK SRIDYNHPGR VLLQNLTMSY SGLYQCTAGN EAGKESCVVR VTVQYVQSIG MVA
Videos
SDS
Documents
Certificates