missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CNKSR2 Protein
A DNA sequence encoding the Homo sapiens (Human) CNKSR2, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_001162118.1 |
---|---|
Gene ID (Entrez) | 22866 |
Name | CNKSR2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16097561
|
enQuireBio™
QP9073-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037571
|
enQuireBio™
QP9073-YE-500UG |
500 μg |
1665.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007571
|
enQuireBio™
QP9073-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027571
|
enQuireBio™
QP9073-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16047571
|
enQuireBio™
QP9073-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017571
|
enQuireBio™
QP9073-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001162118.1 | |
CNKSR2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
22866 | |
Research Use Only | |
CNKSR2 | |
Recombinant Protein | |
Yeast | |
AAEHLDDMNR WLNRINMLTA GYAERERIKQ EQDYWSESDK EEADTPSTPK QDSPPPPYDT YPRPPSMSCA SPYVEAKHSR LSSTETSQSQ SSHEEFRQEV TGSSAVSPIR KTASQRRSWQ DLIETPLTSS GLHYLQTLPL EDSVFSDSAA I | |
Greater than 90% as determined by SDS-PAGE. |