missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CNPY3 Protein

A cDNA sequence encoding the CNPY3 was constructed and used to recombinantly synthesize the protein.

131.00 € - 3915.00 €

Specifications

Name CNPY3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15907368
View Documents Promotion Details
enQuireBio™
QP11470-5UG
5 μg
131.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15997358
View Documents Promotion Details
enQuireBio™
QP11470-20UG
20 μg
203.00 €
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15987358
View Documents Promotion Details
enQuireBio™
QP11470-1MG
1 mg
3915.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CNPY3 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CNPY3 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSGPSQAGA EENDWVRLPS KCEVCKYVAV ELKSAFEETG KTKEVIGTGY GILDQKASGV KYTKSDLRLI EVTETICKRL LDYSLHKERT GSNRFAKGMS ETFETLHNLV HKGVKVVMDI PYELWNETSA EVADLKKQCD VLVEEFEEVI EDWYRNHQEE DLTEFLCANH VLKGKDTSCL AEQWSGKKGD TAALGGKKSK KKSSRAKAAG GRSSSSKQRK ELGGLEGDPS PEEDEGIQKA SPLTHSPPDE L
Greater than 85% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications