missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coagulation Factor II / FII / F2 Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation Factor II / FII / F2, was expressed in the hosts and tags indicated.
287.00 € - 3175.00 €
Specifications
Accession Number | NP_000497.1 |
---|---|
Gene ID (Entrez) | 2147 |
Name | Coagulation Factor II / FII / F2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15919339
|
enQuireBio™
QP6009-YE-100UG |
100 μg |
880.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959339
|
enQuireBio™
QP6009-YE-500UG |
500 μg |
2535.00 €
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929339
|
enQuireBio™
QP6009-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949339
|
enQuireBio™
QP6009-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15969339
|
enQuireBio™
QP6009-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939339
|
enQuireBio™
QP6009-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000497.1 | |
Coagulation Factor II / FII / F2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2147 | |
Research Use Only | |
F2 | |
Recombinant Protein | |
Yeast | |
ANTFLEEVRK GNLERECVEE TCSYEEAFEA LESSTATDVF WAKYTACETA RTPRDKLAAC LEGNCAEGLG TNYRGHVNIT RSGIECQLWR SRYPHKPEIN STTHPGADLQ ENFCRNPDSS TTGPWCYTTD PTVRRQECSI PVCGQDQVTV AMTPRSEGSS VNLSPPLEQC VPDRGQQYQG RLAVTTHGLP CLAWASAQAK ALSKHQDFNS AVQLVENFCR NPDGDEEGVW CYVAGKPGDF GYCDLNYCEE AVEEETGDGL DEDSDRAIEG RTATSEYQTF FNPRTFGSGE ADCGLRPLFE KKSLEDKTER ELLESYIDGR IVEGSDAEIG MSPWQVMLFR KSPQELLCGA SLISDRWVLT AAHCLLYPPW DKNFTENDLL VRIGKHSRTR YERNIEKISM LEKIYIHPRY NWRENLDRDI ALMKLKKPVA FSDYIHPVCL PDRETAASLL QAGYKGRVTG WGNLKETWTA NVGKGQPSVL QVVNLPIVER PVCKDSTRIR ITDNMFCAGY KPDEGKRGDA CEGDSGGPFV MKSPFNNRWY QMGIVSWGEG CDRDGKYGFY THVFRLKKWI QKVIDQFGE | |
Greater than 90% as determined by SDS-PAGE. |