missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(IV) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(IV) chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5852-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001290039.1 | |
Collagen alpha-1(IV) chain Protein | |
Research Use Only | |
COL4A1 | |
Recombinant Protein | |
E. coli | |
GFLVTRHSQT IDDPQCPSGT KILYHGYSLL YVQGNERAHG QDLGTAGSCL RKFSTMPFLF CNINNVCNFA SRNDYSYWLS TPEPMPMSMA PITGENIRPF ISRCAVCEAP AMVMAVHSQT IQIPPCPSGW SSLWIGYSFV MHTSAGAEGS GQALASPGSC LEEFRSAPFI ECHGRGTCNY YANAYSFWLA TIERSEMFKK PTPSTLKAGE LRTHVSRCQV CMRRT | |
Greater than 90% as determined by SDS-PAGE. |
1282 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |