missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XVIII) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XVIII) chain, was expressed in the hosts and tags indicated.
202.00 € - 2645.00 €
Specifications
Accession Number | NP_085059.2 |
---|---|
Gene ID (Entrez) | 80781 |
Name | Collagen alpha-1(XVIII) chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026281
|
enQuireBio™
QP8749-YE-100UG |
100 μg |
615.00 €
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16046281
|
enQuireBio™
QP8749-YE-1MG |
1 mg |
2645.00 €
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16036281
|
enQuireBio™
QP8749-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056281
|
enQuireBio™
QP8749-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066281
|
enQuireBio™
QP8749-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076281
|
enQuireBio™
QP8749-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_085059.2 | |
Collagen alpha-1(XVIII) chain Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
80781 | |
Research Use Only | |
COL18A1 | |
Recombinant Protein | |
Yeast | |
QPVLHLVALN SPLSGGMRGI RGADFQCFQQ ARAVGLAGTF RAFLSSRLQD LYSIVRRADR AAVPIVNLKD ELLFPSWEAL FSGSEGPLKP GARIFSFDGK DVLRHPTWPQ KSVWHGSDPN GRRLTESYCE TWRTEAPSAT GQASSLLGGR LLGQSAASCH HAYIVLCIEN SFMTASK | |
Greater than 90% as determined by SDS-PAGE. |