missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COMP Protein
A cDNA sequence encoding the COMP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11483-1mg
Specifications
COMP Protein | |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
HVDYKDDDDK PAGQGQSPLG SDLGPQMLRE LQETNAALQD VRELLRQQVR EITFLKNTVM ECDACGMQQS VRTGLPSVRP LLHCAPGFCF PGVACIQTES GARCGPCPAG FTGNGSHCTD VNECNAHPCF PRVRCINTSP GFRCEACPPG YSGPTHQGVG LAFAKANKQV CTDINECETG QHNCVPNSVC INTRGSFQCG PCQPGFVGDQ ASGCQRRAQR FCPDGSPSEC HEHADCVLER DGSRSCVCAV GWAGNGILCG RDTDLDGFPD EKLRCPERQC RKDNCVTVPN SGQEDVDRDG IGDACDPDAD GDGVPNEKDN CPLVRNPDQR NTDEDKWGDA CDNCRSQKND DQKDTDQDGR GDACDDDIDG DRIRNQADNC PRVPNSDQKD SDGDGIGDAC DNCPQKSNPD QADVDHDFVG DACDSDQDQD GDGHQDSRDN CPTVPNSAQE DSDHDGQGDA CDDDDDNDGV PDSRDNCRLV PNPGQEDADR DGVGDVCQDD FDADKVVDKI DVCPENAEVT LTDFRAFQTV VLDPEGDAQI DPNWVVLNQG REIVQTMNSD PGLAVGYTAF NGVDFEGTFH VNTVTDDDYA GFIFGYQDSS SFYVVMWKQM EQTYWQANPF RAVAEPGIQL KAVKSSTGPG EQLRNALWHT GDTESQVRLL WKDPRNVGWK DKKSYRWFLQ HRPQVGYIRV RFYEGPELVA DSNVVLDTTM RGGRLGVFCF SQENIIWANL RYRCNDTIPE DYETHQLRQA |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
COMP HEK Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS and 50mM NaCl, pH 7.5 |