missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COX5B Protein
A cDNA sequence encoding the COX5B was constructed and used to recombinantly synthesize the protein.
131.00 € - 1740.00 €
Specifications
Gene ID (Entrez) | 1329 |
---|---|
Name | COX5B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | COX5B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15967428
|
enQuireBio™
QP11492-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15957428
|
enQuireBio™
QP11492-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947428
|
enQuireBio™
QP11492-100UG |
100 μg |
1740.00 €
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
1329 | |
Research Use Only | |
COX5B | |
Human | |
His | |
COX5B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 20% glycerol and 1mM DTT. |
COX5B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSASGGGVP TDEEQATGLE REIMLAAKKG LDPYNVLAPK GASGTREDPN LVPSISNKRI VGCICEEDNT SVVWFWLHKG EAQRCPRCGA HYKLVPQQLA H | |
Greater than 95.0% as determined by SDS-PAGE. |