missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CPEB1 Protein

A DNA sequence encoding the Homo sapiens (Human) CPEB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8045-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042351

  • 518.00 € / 100µg
Estimated Shipment: 02-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001073001.1
CPEB1 Protein
100 μg
Not determined.
Full Length of isoform 4
Human
His-SUMO
Tris-based buffer, 50% glycerol
64506
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CPEB1
Recombinant Protein
E. coli
MLFPTSAQES SRGLPDANDL CLGLQSLSLT GWDRPWSTQD SDSSAQSSTH SVLSMLHNPL GNVLGKPPLS FLPLDPLGSD LVDKFPAPSV RGSRLDTRPI LDSRSSSPSD SDTSGFSSGS DHLSDLISSL RISPPLPFLS LSGGGPRDPL KMGVGSRMDQ EQAALAAVTP SPTSASKRWP GASVWPSWDL LEAPKDPFSI EREARLHRQA AAVNEATCTW SGQLPPRNYK NPIYSCKVFL GGVPWDITEA GLVNTFRVFG SLSVEWPGKD GKHPRCPPKG YVYLVFELEK SVRSLLQACS HDPLSPDGLS EYYFKMSSRR MRCKEVQVIP WVLADSNFVR SPSQRLDPSR TVFVGALHGM LNAEALAAIL NDLFGGVVYA GIDTDKHKYP IGSGRVTFNN QRSYLKAVSA AFVEIKTTKF TKKVQIDPYL EDSLCHICSS QPGPFFCRDQ VCFKYFCRSC WHWRHSMEGL RHHSPLMRNQ KNRDSS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers