missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CPSF4 Protein

A DNA sequence encoding the Homo sapiens (Human) CPSF4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5858-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15968279

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001075028.1
CPSF4 Protein
Research Use Only
CPSF4
Recombinant Protein
E. coli
MQEIIASVDH IKFDLEIAVE QQLGAQPLPF PGMDKSGAAV CEFFLKAACG KGGMCPFRHI SGEKTVVCKH WLRGLCKKGD QCEFLHEYDM TKMPECYFYS KFGECSNKEC PFLHIDPESK IKDCPWYDRG FCKHGPLCRH RHTRRVICVN YLVGFCPEGP SCKFMHPRFE LPMGTTEQPP LPQQTQPPAK QRTPQVIGVM QSQNSSAGNR GPRPLEQVTC YKCGEKGHYA NRCTKGHLAF LSGQ
Greater than 80% as determined by SDS-PAGE.
10898
500 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers