missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CREBZF Protein
A cDNA sequence encoding the CREBZF was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | CREBZF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15907458
|
enQuireBio™
QP11500-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15987448
|
enQuireBio™
QP11500-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15997448
|
enQuireBio™
QP11500-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CREBZF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CREBZF protein solution (0.25 mg/ml) containing PBS buffer (pH 7.4), 20% glycerol, 1mM DTT and 0.1mM PMSF. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMRHSLTK LLAASGSNSP TRSESPEPAA TCSLPSDLTR AAAGEEETAA AGSPGRKQQF GDEGELEAGR GSRGGVAVRA PSPEEMEEEA IASLPGEETE DMDFLSGLEL ADLLDPRQPD WHLDPGLSSP GPLSSSGGGS DSGGLWRGDD DDEAAAAEMQ RFSDLLQRLL NGIGGCSSSS DSGSAEKRRR KSPGGGGGGG SGNDNNQAAT KSPRKAAAAA ARLNRLKKKE YVMGLESRVR GLAAENQELR AENRELGKRV QALQEESRYL RAVLANETGL ARLLSRLSGV GLRLTTSLFR DSPAGDHDYA LPVGKQKQDL LEEDDSAGGV CLHVDKDKVS VEFCSACARK ASSSLKM | |
Greater than 85% as determined by SDS-PAGE. |