missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CREBZF Protein

A cDNA sequence encoding the CREBZF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11500-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987448

Quantity Price
1 171.64 € / 10µg
10 161.27 € / 10µg
Estimated Shipment: 24-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CREBZF Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CREBZF protein solution (0.25 mg/ml) containing PBS buffer (pH 7.4), 20% glycerol, 1mM DTT and 0.1mM PMSF.
Greater than 85% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMRHSLTK LLAASGSNSP TRSESPEPAA TCSLPSDLTR AAAGEEETAA AGSPGRKQQF GDEGELEAGR GSRGGVAVRA PSPEEMEEEA IASLPGEETE DMDFLSGLEL ADLLDPRQPD WHLDPGLSSP GPLSSSGGGS DSGGLWRGDD DDEAAAAEMQ RFSDLLQRLL NGIGGCSSSS DSGSAEKRRR KSPGGGGGGG SGNDNNQAAT KSPRKAAAAA ARLNRLKKKE YVMGLESRVR GLAAENQELR AENRELGKRV QALQEESRYL RAVLANETGL ARLLSRLSGV GLRLTTSLFR DSPAGDHDYA LPVGKQKQDL LEEDDSAGGV CLHVDKDKVS VEFCSACARK ASSSLKM
Videos
SDS
Documents
Certificates