missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRHBP Protein

A cDNA sequence encoding the CRHBP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11503-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987458

  • 6549.36 € / 1mg

Estimated Shipment: 18-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CRHBP Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS YLELREAADY DPFLLFSANL KRELAGEQPY RRALRCLDML SLQGQFTFTA DRPQLHCAAF FISEPEEFIT IHYDQVSIDC QGGDFLKVFD GWILKGEKFP SSQDHPLPSA ERYIDFCESG LSRRSIRSSQ NVAMIFFRVH EPGNGFTLTI KTDPNLFPCN VISQTPNGKF TLVVPHQHRN CSFSIIYPVV IKISDLTLGH VNGLQLKKSS AGCEGIGDFV ELLGGTGLDP SKMTPLADLC YPFHGPAQMK VGCDNTVVRM VSSGKHVNRV TFEYRQLEPY ELENPNGNSI GEFCLSGL YLELREAADY DPFLLFSANL KRELAGEQPY RRALRCLDML SLQGQFTFTA DRPQLHCAAF FISEPEEFIT IHYDQVSIDC QGGDFLKVFD GWILKGEKFP SSQDHPLPSA ERYIDFCESG LSRRSIRSSQ NVAMIFFRVH EPGNGFTLTI KTDPNLFPCN VISQTPNGKF TLVVPHQHRN CSFSIIYPVV IKISDLTLGH VNGLQLKKSS AGCEGIGDFV ELLGGTGLDP SKMTPLADLC YPFHGPAQMK VGCDNTVVRM VSSGKHVNRV TFEYRQLEPY ELENPNGNSI GEFCLSGL
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CRHBP Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 20mM TRIS and 20mM NaCl, pH 7.5.
Videos
SDS
Documents
Certificates