missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CSF2RB Protein

A cDNA sequence encoding the CSF2RB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11529-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15987538

Quantity Price
1 114.77 € / 2µg
10 107.33 € / 2µg
Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

Specifications

Specifications

CSF2RB Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
CSF2RB protein solution (0.5 mg/ml) containing Phosphate Buffered Saline(pH 7.4) and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Insect
WERSLAGAEE TIPLQTLRCY NDYTSHITCR WADTQDAQRL VNVTLIRRVN EDLLEPVSCD LSDDMPWSAC PHPRCVPRRC VIPCQSFVVT DVDYFSFQPD RPLGTRLTVT LTQHVQPPEP RDLQISTDQD HFLLTWSVAL GSPQSHWLSP GDLEFEVVYK RLQDSWEDAA ILLSNTSQAT LGPEHLMPSS TYVARVRTRL APGSRLSGRP SKWSPEVCWD SQPGDEAQPQ NLECFFDGAA VLSCSWEVRK EVASSVSFGL FYKPSPDAGE EECSPVLREG LGSLHTRHHC QIPVPDPATH GQYIVSVQPR RAEKHIKSSV NIQMAPPSLN VTKDGDSYSL RWETMKMRYE HIDHTFEIQY RKDTATWKDS KTETLQNAHS MALPALEPST RYWARVRVRT SRTGYNGIWS EWSEARSWDT ESVLPMWLEH HHHHH
Videos
SDS
Documents
Certificates