missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL16 Protein
A cDNA sequence encoding the CXCL16 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Gene ID (Entrez) | 58191 |
---|---|
Name | CXCL16 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | CXCL16 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15904278
|
enQuireBio™
QP10568-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994268
|
enQuireBio™
QP10568-25UG |
25 μg |
203.00 €
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984268
|
enQuireBio™
QP10568-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
58191 | |
Research Use Only | |
CXCL16 | |
Recombinant Protein | |
E. coli | |
NEGSVTGSCY CGKRISSDSP PSVQFMNRLR KHLRAYHRCL YYTRFQLLSW SVCGGNKDPW VQELMSCLDL KECGHAYSGI VAHQKHLLP | |
Greater than 97.0% as determined by SDS-PAGE. |
CXCL16 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50, as measured by its ability to chemoattract mouse CXCR6-transfected mouse BaF3 cells, is less than 12ng/ml. | |
Human | |
Untagged | |
CXCL16 protein was lyophilized from a 0.2microm filtered solution in PBS. |