missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CXCL16 Protein

A cDNA sequence encoding the CXCL16 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10568-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984268

  • 3401.86 € / 1mg

Estimated Shipment: 23-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

58191
Greater than 97.0% as determined by SDS-PAGE.
Research Use Only
CXCL16
Recombinant Protein
E. coli
NEGSVTGSCY CGKRISSDSP PSVQFMNRLR KHLRAYHRCL YYTRFQLLSW SVCGGNKDPW VQELMSCLDL KECGHAYSGI VAHQKHLLP
CXCL16 Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50, as measured by its ability to chemoattract mouse CXCR6-transfected mouse BaF3 cells, is less than 12ng/ml.
Human
Untagged
CXCL16 protein was lyophilized from a 0.2microm filtered solution in PBS.
Videos
SDS
Documents
Certificates